
Proquinazid

Synonym(s):
6-Iodo-2-propoxy-3-propyl-4(3H)quinazolinone
Empirical Formula (Hill Notation):
C14H17IN2O2
CAS Number:
Molecular Weight:
372.20
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2