
Propofol EP Impurity C

Synonym(s):
2-Isopropylphenol
Linear Formula:
(CH3)2CHC6H4OH
CAS Number:
Molecular Weight:
136.19
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing