
Polysorbate 40

Synonym(s):
Polyethylene glycol sorbitan monopalmitate, Polyoxyethylene sorbitanmonopalmitate
CAS Number:
Products
Brand
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2