
Palmitic acid-1-13C

Synonym(s):
Hexadecanoic acid-1-13C
Linear Formula:
CH3(CH2)1413CO2H
CAS Number:
Molecular Weight:
257.42
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing