
Nitric Oxide Synthase, Inducible from mouse

Synonym(s):
Inducible Nitric Oxide Synthase, NOS II, iNOS, macNOS
Products
Product Category
Brand
Gene Alias
Physical Form
Recombinant Host
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing