![N-Debenzoyl-N-tigloyl-7-epi-paclitaxel certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/907/621/0d80af8f-bb0b-422b-968f-442cc3ce150d/640/0d80af8f-bb0b-422b-968f-442cc3ce150d.png)
N-Debenzoyl-N-tigloyl-7-epi-paclitaxel
![N-Debenzoyl-N-tigloyl-7-epi-paclitaxel certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/907/621/0d80af8f-bb0b-422b-968f-442cc3ce150d/640/0d80af8f-bb0b-422b-968f-442cc3ce150d.png)
Synonym(s):
7-epi-Cephalomannine
Empirical Formula (Hill Notation):
C45H53NO14
CAS Number:
Molecular Weight:
831.90
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing