
N4-Acetylsulfamethoxazole

Synonym(s):
N-{4-{[(5-Methyl-3-isoxazolyl)amino]sulfonyl}phenyl}acetamide, Sulfisomezole-N4-acetate
Empirical Formula (Hill Notation):
C12H13N3O4S
CAS Number:
Molecular Weight:
295.31
Products
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing