Skip to Content
Merck
Minoxidil British Pharmacopoeia (BP) Reference Standard

Minoxidil

Minoxidil British Pharmacopoeia (BP) Reference Standard
Synonym(s):
6-(1-Piperidinyl)-2,4-pyrimidinediamine 3-oxide, 6-(1-Piperidinyl)pyrimidine-2,4-diamine 3-oxide
Empirical Formula (Hill Notation):
C9H15N5O
CAS Number:
Molecular Weight:
209.25
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Application
API Family
Reference Material Type
Special Grade