
Minimum Essential Medium Eagle, with Earl's salts

Synonym(s):
Minimum Essential Medium Eagle, MEM
Products
Brand
Physical Form
Earles Salts
Glucose (g/L)
Hanks Salts
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Sodium Pyruvate (g/L)
Stable Glutamine
Quality Segment
Sterilization Method
Technique
Showing 1-1 of 1