
Meradimate

Synonym(s):
Menthyl anthranilate, Menthyl o-aminobenzoate
Empirical Formula (Hill Notation):
C17H25NO2
CAS Number:
Molecular Weight:
275.39
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing