
Mefloquine hydrochloride

Synonym(s):
(AS)-rel-a-(2R)-2-Piperidinyl-2,8-bis(trifluoromethyl)-4-quinolinemethanol monohydrochloride, Mefloquine hydrochloride
Empirical Formula (Hill Notation):
C17H17ClF6N2O
CAS Number:
Molecular Weight:
414.77
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing