
Lycorine hydrochloride

Empirical Formula (Hill Notation):
C16H17NO4 · HCl
CAS Number:
Molecular Weight:
323.77
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing