
L-Tyrosine-(phenyl-13C6)

Synonym(s):
L-4-Hydroxyphenyl-13C6-alanine
Linear Formula:
4-(HO)13C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
187.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing