
Loperamide for system suitability

Synonym(s):
Loperamide hydrochloride, 4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutyramide hydrochloride
Empirical Formula (Hill Notation):
C29H33ClN2O2 · HCl
CAS Number:
Molecular Weight:
513.50
Products
Formula Weight
Gene Alias
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing