
L-Arginine-13C6 hydrochloride

Synonym(s):
(S)-(+)-2-Amino-5-[(aminomethyl)amino]pentanoic acid-13C6 hydrochloride, 13C Labeled arginine, 13C Labeled arginine hydrochloride
Linear Formula:
H2N13C(NH)NH13CH2(13CH2)213CH(NH2)13CO2H · HCl
CAS Number:
Molecular Weight:
216.62
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing