
Isomenthone

Synonym(s):
cis-2-Isopropyl-5-methylcyclohexanone, cis-p-Menthan 3-one, cis-Menthone
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing