Skip to Content
Merck
Iron(III) acetylacetonate purum, ≥97.0% (RT)

Iron(III) acetylacetonate

Iron(III) acetylacetonate purum, ≥97.0% (RT)
Synonym(s):
2,4-Pentanedione iron(III) derivative, Fe(acac)3, Ferric acetylacetonate, Iron(III) 2,4-pentanedionate
Linear Formula:
Fe(C5H7O2)3
CAS Number:
Molecular Weight:
353.17
EC Number:

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-3 of 3