
Imiprothrin

Synonym(s):
2-Propargylsuccinimidylmethyl chrysanthemate
Empirical Formula (Hill Notation):
C17H22N2O4
CAS Number:
Molecular Weight:
318.37
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing