
Hexagonal boron nitride ink

Empirical Formula (Hill Notation):
BN
CAS Number:
Molecular Weight:
24.82
Products
Brand
Sustainability
Boiling Point (°C)
Color
Feature
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing