
Guaiacol Impurity C

Synonym(s):
1,2-Dimethoxybenzene, Pyrocatechol dimethyl ether, Veratrole
Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing