
Ginsenoside Rk1

Synonym(s):
(3β, 12β)-12-Hydroxydammara-20,24-dien-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C42H70O12
CAS Number:
Molecular Weight:
767.00
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing