
FTY720

Synonym(s):
2-Amino-2-[2-(4-octyl-phenyl)-ethyl]-propane-1,3-diol hydrochloride, Fingolimod hydrochloride
Empirical Formula (Hill Notation):
C19H33NO2 · HCl
CAS Number:
Molecular Weight:
343.93
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing