
Flucytosine

Synonym(s):
5-Fluorocytosine, 4-amino-5-fluoro-2(1H)-pyrimidinone, Flucytosine
Empirical Formula (Hill Notation):
C4H4FN3O
CAS Number:
Molecular Weight:
129.09
Products
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique