
Famoxadone

Synonym(s):
3-Anilino-5-methyl-5-(4-phenoxyphenyl)oxazolidine-2,4-dione
Empirical Formula (Hill Notation):
C22H18N2O4
CAS Number:
Molecular Weight:
374.39
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing