
EX-CELL® 302 Serum-Free Medium for CHO Cells

Products
Product Category
Brand
Physical Form
Product Line
Glucose (g/L)
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Sterilization Method
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing