
Efavirenz Related Compound C

Synonym(s):
6-Chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline
Empirical Formula (Hill Notation):
C13H9ClF3N
CAS Number:
Molecular Weight:
271.67
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2