
(E)-10-Hydroxy amitriptyline

Synonym(s):
Amitriptyline metabolite, (±)-E-10-hydroxylated-, (±)-(E)-5-[3-(Dimethylamino)propylidene]-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-10-ol
Empirical Formula (Hill Notation):
C20H23NO
CAS Number:
Molecular Weight:
293.40
Products
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing