
DTT

Synonym(s):
DL-Dithiothreitol, threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT
Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing