
Diazepam for system suitability

Synonym(s):
Diazepam, 7-Chloro-1-methyl-5-phenyl-3H-1,4-benzodiazepin-2(1H)-one, Ro 5-2807
Empirical Formula (Hill Notation):
C16H13ClN2O
CAS Number:
Molecular Weight:
284.74
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing