
Clarithromycin

Empirical Formula (Hill Notation):
C38H69NO13
CAS Number:
Molecular Weight:
747.95
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing