
Cefamandole nafate

Synonym(s):
Cefamandole formate sodium salt
Linear Formula:
C19H17N6O6S2Na
CAS Number:
Molecular Weight:
512.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing