
Bromperidol

Synonym(s):
4-[4-(4-Bromophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, R 11333
Empirical Formula (Hill Notation):
C21H23BrFNO2
CAS Number:
Molecular Weight:
420.32
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing