Skip to Content
KH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Barium chloride
Linear Formula:
BaCl
2
CAS Number:
10361-37-2
Molecular Weight:
208.23
EC Number:
233-788-1
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Anhydrous (2)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
342920
Barium chloride,
99.9% trace metals basis
Expand
View Pricing
202738
Barium chloride,
99.999% trace metals basis
Expand
View Pricing
449644
Barium chloride,
AnhydroBeads
™
, −10 mesh, 99.999% trace metals basis
Expand
View Pricing
449652
Barium chloride,
AnhydroBeads
™
, −10 mesh, 99.95% trace metals basis
Expand
View Pricing