
Atomoxetine Related Compound B

Synonym(s):
N-Methyl-γ-(3-methylphenoxy)benzenepropanamine hydrochloride, N-Methyl-3-phenyl-3-(3-methylphenoxy)propylamine hydrochloride
Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2