
Adapalene Related Compound E

Synonym(s):
2,2′-Binaphthyl-6,6′-dicarboxylic acid, 2,2′-Binaphthalene-6,6′-dicarboxylic acid
Empirical Formula (Hill Notation):
C22H14O4
CAS Number:
Molecular Weight:
342.34
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2