
6-Hydroxy-DL-DOPA

Synonym(s):
2,4,5-Trihydroxy-DL-phenylalanine, 2,5-Dihydroxy-DL-tyrosine
Empirical Formula (Hill Notation):
C9H11NO5
CAS Number:
Molecular Weight:
213.19
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing