![4-Amino-3-iodobenzonitrile 98%](/deepweb/assets/sigmaaldrich/product/structures/362/660/591c33cd-2ccd-4816-9205-d8daf6a2e529/640/591c33cd-2ccd-4816-9205-d8daf6a2e529.png)
4-Amino-3-iodobenzonitrile
![4-Amino-3-iodobenzonitrile 98%](/deepweb/assets/sigmaaldrich/product/structures/362/660/591c33cd-2ccd-4816-9205-d8daf6a2e529/640/591c33cd-2ccd-4816-9205-d8daf6a2e529.png)
Synonym(s):
1-Amino-2-iodo-4-cyanobenzene, 4-Cyano-2-iodoaniline
Linear Formula:
IC6H3(NH2)CN
CAS Number:
Molecular Weight:
244.03
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing