Skip to Content
Merck
3-Pentanol 98%

3-Pentanol

3-Pentanol 98%
Synonym(s):
sec-inact.-Pentyl alcohol, Diethyl carbinol
Linear Formula:
(C2H5)2CHOH
CAS Number:
Molecular Weight:
88.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing