
3-(4-methoxyphenyl)pyrazole

Empirical Formula (Hill Notation):
C10H10N2O
CAS Number:
Molecular Weight:
174.20
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing