
19-Norethindrone

Synonym(s):
17α-Ethynyl-19-nortestosterone, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one, 17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one, 19-Nor-17α-ethynyl-4-androsten-17β-ol-3-one, 19-Norethisterone
Empirical Formula (Hill Notation):
C20H26O2
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing