
1,1,1-Tris(diphenylphosphinomethyl)ethane

Synonym(s):
1,3-Bis(diphenylphosphino)-2-(diphenylphosphino)methyl-2-methylpropane, Triphos
Linear Formula:
CH3C[CH2P(C6H5)2]3
CAS Number:
Molecular Weight:
624.67
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing