Mobile Phase Optimization in the Analysis of Lysozyme on Zenix® SEC-300
Materials
analytical column
Product No.
Description
Pricing
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 4.6 mm, 3 μm particle size, stainless steelCONDITIONS
column
Zenix SEC-300, 30 cm x 4.6 mm I.D., 3 μm particles (Z777028)
mobile phase
as indicated on figure
flow rate
0.35 mL/min
temp.
25 °C
detector
UV, 214 nm
injection
5 μL
sample
lysozyme, 1 mg/mL
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies