Merck
Population genetic structure of the endemic rosewoods
Ecology and evolution (2018-01-13)
Ida Hartvig, Thea So, Suchitra Changtragoon, Hoa Thi Tran, Somsanith Bouamanivong, Ida Theilade, Erik Dahl Kjær, Lene Rostgaard Nielsen
ABSTRACT
Indochina is a biodiversity hot spot and harbors a high number of endemic species, most of which are poorly studied. This study explores the genetic structure and reproductive system of the threatened endemic timber species
Sigma-Aldrich
2-Methyl-2-propanethiol, 99%