Merck
  • The fetal ovary exhibits temporal sensitivity to a 'real-life' mixture of environmental chemicals.
The fetal ovary exhibits temporal sensitivity to a 'real-life' mixture of environmental chemicals.
Scientific reports (2016-03-05)
Richard G Lea, Maria R Amezaga, Benoit Loup, Béatrice Mandon-Pépin, Agnes Stefansdottir, Panagiotis Filis, Carol Kyle, Zulin Zhang, Ceri Allen, Laura Purdie, Luc Jouneau, Corinne Cotinot, Stewart M Rhind, Kevin D Sinclair, Paul A Fowler
ABSTRACT
The development of fetal ovarian follicles is a critical determinant of adult female reproductive competence. Prolonged exposure to environmental chemicals (ECs) can perturb this process with detrimental consequences for offspring. Here we report on the exposure of pregnant ewes to an environmental mixture of ECs derived from pastures fertilized with sewage sludge (biosolids): a common global agricultural practice. Exposure of pregnant ewes to ECs over 80 day periods during early, mid or late gestation reduced the proportion of healthy early stage fetal follicles comprising the ovarian reserve. Mid and late gestation EC exposures had the most marked effects, disturbing maternal and fetal liver chemical profiles, masculinising fetal anogenital distance and greatly increasing the number of altered fetal ovarian genes and proteins. In conclusion, differential temporal sensitivity of the fetus and its ovaries to EC mixtures has implications for adult ovarian function following adverse exposures during pregnancy.
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-COPS4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution