Merck
  • Total synthesis of macquarimicins using an intramolecular Diels-Alder approach inspired by a biosynthetic pathway.
Total synthesis of macquarimicins using an intramolecular Diels-Alder approach inspired by a biosynthetic pathway.
Journal of the American Chemical Society (2004-09-10)
Ryosuke Munakata, Hironori Katakai, Tatsuo Ueki, Jun Kurosaka, Ken-Ichi Takao, Kin-Ichi Tadano
ABSTRACT
A total synthesis of the macquarimicins A-C (1-3), novel natural products with intriguing tetra- or pentacyclic frameworks, has been achieved. The synthesis features an extensive investigation of the biosynthesis-based intramolecular Diels-Alder (IMDA) reactions of (E,Z,E)-1,6,8-nonatrienes. Considering possible biosynthetic sequences, four types of substrates were synthesized, and their IMDA reactions were examined. From one of the four substrates, the total synthesis was achieved via a transannular Diels-Alder reaction, which led to the stereoselective construction of the unique molecular framework. The convergent and efficient synthetic pathway afforded (+)-1 in 27 linear steps with 4.3% and 9.9% overall yields from readily available ethyl (2E,4S)-4,5-(isopropylidene)dioxy-2-pentenoate (22) and (R)-epichlorohydrin (30), respectively. Furthermore, efficient syntheses of 2, 3, and the 9-epi-cochleamycins A (57) and B (58) were accomplished. Additionally, the present work established the absolute stereochemistry of macquarimicins and revised the C(2)--C(3) geometry of 1.
Sigma-Aldrich
(S)-(+)-Epichlorohydrin, 98%