Merck
  • Elevated frequency of sister chromatid exchanges in lymphocytes of victims of the Tokyo sarin disaster and in experiments exposing lymphocytes to by-products of sarin synthesis.
Elevated frequency of sister chromatid exchanges in lymphocytes of victims of the Tokyo sarin disaster and in experiments exposing lymphocytes to by-products of sarin synthesis.
Toxicology letters (1998-10-17)
Q Li, M Minami, J G Clement, C A Boulet
ABSTRACT
More than 5000 passengers of Tokyo subway trains were injured with toxic chemicals including the nerve gas sarin. Most of the victims examined had marked miosis and decreased serum cholinesterase activity. To monitor the genetic aftereffects of sarin exposure, we measured sister chromatid exchanges (SCEs) of the victims using peripheral blood lymphocytes. The frequency of SCEs was significantly higher in the victims than in the control group. Analyzing results using samples of urine from the victims suggested that the victims were exposed to not only sarin per se, but by-products of sarin synthesis, i.e. diisopropyl methylphosphonate (DIMP), diethyl methylphosphonate (DEMP) and ethyl isopropyl methylphosphonate (EIMP). Thus, the in vitro SCE-inducing effect of DIMP, DEMP and EIMP was examined using human lymphocytes and we obtained positive results.
Sigma-Aldrich
Diethyl methylphosphonate, 97%