Merck
  • Chiral separation and detection enhancement of propranolol using automated pre-column derivatization.
Chiral separation and detection enhancement of propranolol using automated pre-column derivatization.
Journal of pharmaceutical and biomedical analysis (1993-02-01)
F Lai, A Mayer, T Sheehan
ABSTRACT
In the liquid chromatographic analysis of pharmaceuticals, two challenges are often encountered: detectivity and chiral separations. Propranolol, a beta adrenergic blocker, is a pharmaceutical compound that faces both of these limitations. In this study, both limitations are overcome simultaneously using derivatization with (+)-1-(9-fluorenyl)ethyl chloroformate (FLEC), a highly fluorescent and chiral reagent. The derivatization is automated using an autosampler with an AutoMix microrobotic feature, which greatly contributes to the efficiency and reproducibility of the method when manipulating microliter volumes of sample and reagents. The method yields excellent separation of the diasteriomers, has a detection limit of 1 picomol, good reproducibility and linearity in the 50-400 pmol range (on column). In addition, this method is simple, requires no elevated temperature, no chiral stationary or mobile phases and can be easily automated.
Sigma-Aldrich
(+)-1-(9-Fluorenyl)ethyl chloroformate solution, 18 mM in acetone, for chiral derivatization
Supelco
(+)-1-(9-Fluorenyl)ethyl chloroformate solution, ≥18 mM in acetone, for chiral derivatization, LiChropur