Merck
  • Assays of three carcinogen/non-carcinogen chemical pairs for in vivo induction of chromosome aberrations, sister chromatid exchanges and micronuclei.
Assays of three carcinogen/non-carcinogen chemical pairs for in vivo induction of chromosome aberrations, sister chromatid exchanges and micronuclei.
Environmental and molecular mutagenesis (1989-01-01)
A F McFee, P P Jauhar, K W Lowe, J T MacGregor, C M Wehr
ABSTRACT
Three pairs of structurally similar carcinogenic/non-carcinogenic chemicals were tested for in vivo genotoxic activity in B6C3F1 mice. The carcinogenic/non-carcinogenic pairs, respectively, were o-toluidine hydrochloride/o-anthranilic acid, 4-chloro-o-phenylenediamine/4-nitro-o-phenylenediamine, and 3-(chloromethyl)pyridine hydrochloride/2-(chloromethyl)pyridine hydrochloride. Bone marrow cells from mice given intraperitoneal injections of up to the maximum tolerated dose were evaluated for chromosomal aberration, sister chromatid exchange, and micronucleus induction, o-anthranilic acid and o-toluidine hydrochloride did not increase the frequency of chromosomal aberrations or micronuclei. o-Toluidine hydrochloride increased the frequency of sister chromatid exchanges in two successive trials, while o-anthranilic acid had a positive effect on sister chromatid exchanges in two of three trials. Both 2-(chloromethyl) and 3-(chloromethyl)pyridine hydrochloride were negative for all three endpoints. Assays for chromosomal aberrations and micronuclei each distinguished between 4-chloro-o-phenylenediamine and its non-carcinogenic companion, 4-nitro-o-phenylenediamine. In the aberration test, 4-chloro-o-phenylenediamine produced a few cells with very large numbers of aberrations rather than an even distribution of damage among cells.
Sigma-Aldrich
4-Chloro-o-phenylenediamine, 97%
Sigma-Aldrich
3-(Chloromethyl)pyridine hydrochloride, 96%