Merck
  • Study of broadband near-infrared emission in Tm3+-Er3+ codoped TeO2-WO3-PbO glasses.
Study of broadband near-infrared emission in Tm3+-Er3+ codoped TeO2-WO3-PbO glasses.
Optics express (2009-05-26)
R Balda, J Fernández, J M Fernández-Navarro
ABSTRACT
In this work, we report the near-infrared emission properties of Tm(3+)-Er(3+) codoped tellurite TeO(2)-WO(3)-PbO glasses under 794 nm excitation. A broad emission from 1350 to 1750 nm corresponding to the Tm(3+) and Er(3+) emissions is observed. The full width at half-maximum of this broadband increases with increasing [Tm]/[Er] concentration ratio up to a value of ~ 160 nm. The energy transfer between Tm(3+) and Er(3+) ions is evidenced by both the temporal behavior of the near-infrared luminescence and the effect of Tm3+ codoping on the visible upconversion of Er(3+) ions.
Sigma-Aldrich
Tellurium dioxide, 99.995% trace metals basis
Sigma-Aldrich
Tellurium dioxide, ≥99%
Sigma-Aldrich
Tellurium dioxide, ≥97.0%