Merck
  • Haemopoietic reconstitution after sublethal irradiation: comparison of the effects of different haemopoietic cytokines on murine lymphocytes and bone marrow cells.
Haemopoietic reconstitution after sublethal irradiation: comparison of the effects of different haemopoietic cytokines on murine lymphocytes and bone marrow cells.
British journal of haematology (1999-08-25)
D Frasca, F Guidi, P Barattini, G Doria
ABSTRACT
Sublethally irradiated mice were injected with recombinant cytokines to stimulate haemopoietic reconstitution. Interleukin (IL)-11 and IL-6 were able to significantly accelerate the recovery of thymus, spleen and bone marrow cells when used in combination with IL-3, but not alone. Stem cell factor (SCF) also displayed detectable effects when used with IL-3. Conversely, the IL-6 superagonist K-7/D-6 was able, even when used alone, to induce recovery of thymus, spleen and bone marrow cells up to the level of unirradiated controls. Together, these results indicate that it is possible to attain complete recovery of lymphoid organs and tissues as early as 7 d after irradiation by use of haemopoietic cytokines.
Sigma-Aldrich
Casein from bovine milk, technical grade
Sigma-Aldrich
Interferon-γ Protein, Recombinant human, The Interferon-g protein (or IFN-gamma protein) is a lymphoid factor which possesses potent anti-viral activity.